Learn More
Grainger Bumper 1/2 in Black Rubber PK50
Catalog No. 502764819
Encompass_Preferred
The Grainger 16WD13 Bumper Door Stop is a door-mounted door stop made of rubber and steel. It has a diameter of 1-1/4", projects 2-1/2" from the door surface, and withstands up to 100 lbs of force. This door stop features a spring-loaded plunger to absorb shock and reduce noise. It can be mounted with screws (not included) and is designed for use on wood or metal doors. The door stop helps protect walls and door handles by cushioning the impact when doors are opened forcefully. It comes in black color.
Catalog No. 50-276-4819 Supplier Grainger Supplier No. 16WD13
The Grainger 16WD13 Bumper Door Stop is a door-mounted door stop made of rubber and steel. It has a diameter of 1-1/4", projects 2-1/2" from the door surface, and withstands up to 100 lbs of force. This door stop features a spring-loaded plunger to absorb shock and reduce noise. It can be mounted with screws (not included) and is designed for use on wood or metal doors. The door stop helps protect walls and door handles by cushioning the impact when doors are opened forcefully. It comes in black color.