Learn More
Grainger Bumper 1/2 in Black Rubber PK50
Catalog No. 502764723
Encompass_Preferred
The Grainger 1/2" Black Rubber Bumper is a rubber bumper with a 1/2" diameter and 1-1/4" length. It has a hole diameter of 1/4" and is made of SBR rubber. This bumper absorbs shock and reduces noise from vibration. It can be used on bins, carts, conveyors, doors, drawers, and more. The operating temperature range is -20°F to +180°F. It meets military specification MIL-P-15011C. This bumper comes in a convenient pack of 50 pieces.
Catalog No. 50-276-4723 Supplier Grainger Supplier No. 16WC41
The Grainger 1/2" Black Rubber Bumper is a rubber bumper with a 1/2" diameter and 1-1/4" length. It has a hole diameter of 1/4" and is made of SBR rubber. This bumper absorbs shock and reduces noise from vibration. It can be used on bins, carts, conveyors, doors, drawers, and more. The operating temperature range is -20°F to +180°F. It meets military specification MIL-P-15011C. This bumper comes in a convenient pack of 50 pieces.