Learn More
Grainger Bumper 1 in Black Rubber PK10
Catalog No. 502767762
Encompass_Preferred
The 16ZD48 is a 3M Scotch-Brite General Purpose Hand Pad, 6" x 9", from Grainger. It features an open web construction that cleans effectively without scratching. The pad is made of high-quality fibers and resin that resist shredding and matting. It can be used for scrubbing pots, pans, dishes, floors, walls, and other surfaces. The pad measures 6 inches by 9 inches and has a maximum use temperature of 220°F. It is disposable and ideal for light-duty scrubbing.
Catalog No. 50-276-7762 Supplier Grainger Supplier No. 16ZD48
The 16ZD48 is a 3M Scotch-Brite General Purpose Hand Pad, 6" x 9", from Grainger. It features an open web construction that cleans effectively without scratching. The pad is made of high-quality fibers and resin that resist shredding and matting. It can be used for scrubbing pots, pans, dishes, floors, walls, and other surfaces. The pad measures 6 inches by 9 inches and has a maximum use temperature of 220°F. It is disposable and ideal for light-duty scrubbing.