Learn More
Grainger Bumper 1 in Black Rubber PK10
Catalog No. 502768135
Encompass_Preferred
The Grainger Approved 1" Black Rubber Bumper is a rubber bumper with a 1" diameter and 1" height. It has a low durometer for shock absorption and is made of oil and water resistant neoprene rubber. The bumper has a pressure-sensitive adhesive backing for easy installation. It can be used as a rubber foot to reduce vibration, a non-skid bumper, or for spacing and cushioning. The operating temperature range is -20°F to 200°F. This versatile rubber bumper comes in packs of 4 or 8.
Catalog No. 50-276-8135 Supplier Grainger Supplier No. 16ZD47
The Grainger Approved 1" Black Rubber Bumper is a rubber bumper with a 1" diameter and 1" height. It has a low durometer for shock absorption and is made of oil and water resistant neoprene rubber. The bumper has a pressure-sensitive adhesive backing for easy installation. It can be used as a rubber foot to reduce vibration, a non-skid bumper, or for spacing and cushioning. The operating temperature range is -20°F to 200°F. This versatile rubber bumper comes in packs of 4 or 8.