Learn More
Grainger Bumper 1 in Black Rubber PK10
Catalog No. 502768230
Encompass_Preferred
The 1" Black Rubber Bumper from Grainger is a rubber bumper with a 1" diameter and 1" height. It has a durometer hardness of 60A and is made from SBR rubber. The bumper has a maximum load capacity of 15 lbs and operates in temperatures from -40°F to 180°F. It can be used as a shock absorber or spacer and has a black finish. The bumper comes in a package of 10 pieces.
Catalog No. 50-276-8230 Supplier Grainger Supplier No. 16XU84
The 1" Black Rubber Bumper from Grainger is a rubber bumper with a 1" diameter and 1" height. It has a durometer hardness of 60A and is made from SBR rubber. The bumper has a maximum load capacity of 15 lbs and operates in temperatures from -40°F to 180°F. It can be used as a shock absorber or spacer and has a black finish. The bumper comes in a package of 10 pieces.