Learn More
Grainger Bumper 1 in Black Rubber PK25
Catalog No. 502766498
Encompass_Preferred
The Bumper 1 in Black Rubber by Grainger Industrial Supply is a 1 inch thick black rubber bumper. It is made from SBR rubber which provides shock absorption and has a durometer of 60A. The bumpers have predrilled holes for easy mounting and come in 4 inch, 6 inch, and 8 inch diameters. They can withstand temperatures from -40 to 180 degrees Fahrenheit. These durable rubber bumpers are often used as dock bumpers, corner guards, or to prevent metal to metal contact.
Catalog No. 50-276-6498 Supplier Grainger Supplier No. 16ZD29
The Bumper 1 in Black Rubber by Grainger Industrial Supply is a 1 inch thick black rubber bumper. It is made from SBR rubber which provides shock absorption and has a durometer of 60A. The bumpers have predrilled holes for easy mounting and come in 4 inch, 6 inch, and 8 inch diameters. They can withstand temperatures from -40 to 180 degrees Fahrenheit. These durable rubber bumpers are often used as dock bumpers, corner guards, or to prevent metal to metal contact.