Learn More
Grainger Bumper 11/16 in Black Rubber PK25
Catalog No. 502764218
Encompass_Preferred
The Grainger Approved 16WH21 Bumper is a black rubber bumper with a 2-1/2" projection and a 1-1/2" hole diameter. It is made of SBR rubber and has a durometer of 60A. This bumper provides impact absorption and has a temperature range of -20°F to 180°F. It can be used for material handling equipment, conveyors, and other industrial applications requiring shock absorption. The bumpers are sold individually.
Catalog No. 50-276-4218 Supplier Grainger Supplier No. 16WH21
The Grainger Approved 16WH21 Bumper is a black rubber bumper with a 2-1/2" projection and a 1-1/2" hole diameter. It is made of SBR rubber and has a durometer of 60A. This bumper provides impact absorption and has a temperature range of -20°F to 180°F. It can be used for material handling equipment, conveyors, and other industrial applications requiring shock absorption. The bumpers are sold individually.