Learn More
Grainger Bumper 13/16 in Black Rubber PK10
Catalog No. 502767834
Encompass_Preferred
The Grainger 16XU86 is a Westward 3/8-inch drive impact socket adapter that converts a 3/8-inch drive to a 1/2-inch drive. It is made from chrome vanadium steel for durability and has a matte finish to resist corrosion. This impact socket adapter allows you to use 3/8-inch drive tools with a 1/2-inch drive impact wrench, increasing versatility. It measures 2-1/2 inches long for reach into tight spaces. The 16XU86 socket adapter has a torque rating up to 250 ft-lbs to handle high torque applications.
Catalog No. 50-276-7834 Supplier Grainger Supplier No. 16XU86
The Grainger 16XU86 is a Westward 3/8-inch drive impact socket adapter that converts a 3/8-inch drive to a 1/2-inch drive. It is made from chrome vanadium steel for durability and has a matte finish to resist corrosion. This impact socket adapter allows you to use 3/8-inch drive tools with a 1/2-inch drive impact wrench, increasing versatility. It measures 2-1/2 inches long for reach into tight spaces. The 16XU86 socket adapter has a torque rating up to 250 ft-lbs to handle high torque applications.