Learn More
Grainger Bumper 15/16 in Black Rubber PK10
Catalog No. 502767763
Encompass_Preferred
The Grainger 16XT92 Bumper Door Stop is a door-mounted door stop made of rubber and steel. It has a diameter of 1-1/4", projects 2-1/2" from the door surface, and withstands up to 100 lbs of holding force. This door stop features a mounting bracket that attaches to the door with two included screws. It has a black finish and is designed to protect walls and door surfaces by absorbing the impact of a swinging door. This product is made in the USA and is commonly used in commercial buildings.
Catalog No. 50-276-7763 Supplier Grainger Supplier No. 16XT92
The Grainger 16XT92 Bumper Door Stop is a door-mounted door stop made of rubber and steel. It has a diameter of 1-1/4", projects 2-1/2" from the door surface, and withstands up to 100 lbs of holding force. This door stop features a mounting bracket that attaches to the door with two included screws. It has a black finish and is designed to protect walls and door surfaces by absorbing the impact of a swinging door. This product is made in the USA and is commonly used in commercial buildings.