Learn More
Grainger Bumper 15/16 in Black Rubber PK5
Catalog No. 502768125
Encompass_Preferred
The Grainger 15-16 in Black Rubber Bumper is a rubber bumper with a 15/16 in diameter and a 1-1/2 in face width. It has a 1/4 in mounting hole and is made from SBR rubber with a Shore A durometer hardness of 60A. This industrial bumper absorbs shock and protects equipment from damage. It has excellent weather and abrasion resistance. The operating temperature range is -20 to 180 degrees Fahrenheit.
Catalog No. 50-276-8125 Supplier Grainger Supplier No. 16YR04
The Grainger 15-16 in Black Rubber Bumper is a rubber bumper with a 15/16 in diameter and a 1-1/2 in face width. It has a 1/4 in mounting hole and is made from SBR rubber with a Shore A durometer hardness of 60A. This industrial bumper absorbs shock and protects equipment from damage. It has excellent weather and abrasion resistance. The operating temperature range is -20 to 180 degrees Fahrenheit.