Learn More
Grainger Bumper 15/32 in Black Rubber PK10
Catalog No. 502767967
Encompass_Preferred
The Grainger 16YJ36 Bumper Door Stop is a door-mounted door stop made of rubber and steel. It has a diameter of 1-1/2", projects 2-1/2" from the door surface, and withstands up to 100 lbs of force. This door stop features a low profile bumper design and is designed for high traffic commercial doors. It can be mounted with screws (included) and has pre-drilled holes for easy installation. The door stop is available in black color.
Catalog No. 50-276-7967 Supplier Grainger Supplier No. 16YJ36
The Grainger 16YJ36 Bumper Door Stop is a door-mounted door stop made of rubber and steel. It has a diameter of 1-1/2", projects 2-1/2" from the door surface, and withstands up to 100 lbs of force. This door stop features a low profile bumper design and is designed for high traffic commercial doors. It can be mounted with screws (included) and has pre-drilled holes for easy installation. The door stop is available in black color.