Learn More
Grainger Bumper 3/4 in Black Rubber PK10
Catalog No. 502764371
Encompass_Preferred
The Grainger 3/4" Black Rubber Bumper Washer is a rubber bumper washer designed to help absorb shock and reduce noise. It has an inside diameter of 3/4", an outside diameter of 1-1/4", and is 1/4" thick. This bumper washer is made of oil and water resistant molded rubber and sold in a pack of 10 washers. It can provide cushioning between metal surfaces and is commonly used on doors, lids, and cabinets. This product helps protect surfaces from damage while reducing noise from impact. The black color allows it to blend in seamlessly on most applications.
Catalog No. 50-276-4371 Supplier Grainger Supplier No. 16WC77
The Grainger 3/4" Black Rubber Bumper Washer is a rubber bumper washer designed to help absorb shock and reduce noise. It has an inside diameter of 3/4", an outside diameter of 1-1/4", and is 1/4" thick. This bumper washer is made of oil and water resistant molded rubber and sold in a pack of 10 washers. It can provide cushioning between metal surfaces and is commonly used on doors, lids, and cabinets. This product helps protect surfaces from damage while reducing noise from impact. The black color allows it to blend in seamlessly on most applications.