Learn More
Grainger Bumper 3/4 in Black Rubber PK10
Catalog No. 502764397
Encompass_Preferred
The Grainger 16WJ89 is a Westward 3/8-inch drive impact socket adapter that converts a 3/8-inch drive to a 1/2-inch drive. It is made from chrome vanadium steel for durability and has a matte finish. The adapter measures 2-1/2 inches long with a 3/8 inch female square drive and a 1/2 inch male square drive. It can be used with impact wrenches and is rated up to 20,000 ft-lbs of torque. The adapter allows use of 1/2-inch drive sockets on a 3/8-inch drive tool.
Catalog No. 50-276-4397 Supplier Grainger Supplier No. 16WJ89
The Grainger 16WJ89 is a Westward 3/8-inch drive impact socket adapter that converts a 3/8-inch drive to a 1/2-inch drive. It is made from chrome vanadium steel for durability and has a matte finish. The adapter measures 2-1/2 inches long with a 3/8 inch female square drive and a 1/2 inch male square drive. It can be used with impact wrenches and is rated up to 20,000 ft-lbs of torque. The adapter allows use of 1/2-inch drive sockets on a 3/8-inch drive tool.