Learn More
Grainger Bumper 3/4 in Black Rubber PK25
Catalog No. 502768216
Encompass_Preferred
The 3/4" Black Rubber Bumper from Grainger is a rubber bumper designed to help absorb shock and reduce noise from impacts. It has a 3/4" diameter and is made from durable black rubber. This bumper can be used in a variety of applications including on doors, cabinets, and machinery. It has a temperature range of -20°F to 160°F. The bumper has a durometer of 60A rubber. It meets military specification MIL-B-15011C. The bumper is sold individually.
Catalog No. 50-276-8216 Supplier Grainger Supplier No. 16YF52
The 3/4" Black Rubber Bumper from Grainger is a rubber bumper designed to help absorb shock and reduce noise from impacts. It has a 3/4" diameter and is made from durable black rubber. This bumper can be used in a variety of applications including on doors, cabinets, and machinery. It has a temperature range of -20°F to 160°F. The bumper has a durometer of 60A rubber. It meets military specification MIL-B-15011C. The bumper is sold individually.