Learn More
Grainger Bumper 3/4 in Black Rubber PK25
Catalog No. 502767885
Encompass_Preferred
The 3/4" Black Rubber Bumper from Grainger is a rubber bumper with a 3/4" diameter and 1" length. It is made from black rubber and has a durometer hardness of 60A. The bumpers feature a molded-in steel insert and no holes. They can provide impact absorption and cushioning in machinery and equipment. These rubber bumpers are sold in a convenient pack of 25 pieces.
Catalog No. 50-276-7885 Supplier Grainger Supplier No. 16ZD32
The 3/4" Black Rubber Bumper from Grainger is a rubber bumper with a 3/4" diameter and 1" length. It is made from black rubber and has a durometer hardness of 60A. The bumpers feature a molded-in steel insert and no holes. They can provide impact absorption and cushioning in machinery and equipment. These rubber bumpers are sold in a convenient pack of 25 pieces.