Learn More
Grainger Bumper 3/4 in Black Rubber PK25
Catalog No. 502767966
Encompass_Preferred
The Grainger 16YJ34 is a Westward brand 1/4-inch drive socket set. It contains 15 SAE sockets ranging from 4mm to 14mm and 15 metric sockets ranging from 5/32 inch to 9/16 inch. The chrome vanadium steel sockets have a polished finish and feature a knurled design for easy hand tightening. This 30 piece socket set comes packed in a molded case for convenient storage and portability. The compact case has a hinged lid and socket size markings on the inside for quick identification.
Catalog No. 50-276-7966 Supplier Grainger Supplier No. 16YJ34
The Grainger 16YJ34 is a Westward brand 1/4-inch drive socket set. It contains 15 SAE sockets ranging from 4mm to 14mm and 15 metric sockets ranging from 5/32 inch to 9/16 inch. The chrome vanadium steel sockets have a polished finish and feature a knurled design for easy hand tightening. This 30 piece socket set comes packed in a molded case for convenient storage and portability. The compact case has a hinged lid and socket size markings on the inside for quick identification.