Learn More
Grainger Bumper 3/8 in Black Rubber PK50
Catalog No. 502764817
Encompass_Preferred
The 16WD02 is a 16-inch diameter, 2-blade, direct drive, forward-curved centrifugal fan from Continental Fan Manufacturing. It is constructed of galvanized steel and has a 1/3 HP, 115V, single-phase, permanent split capacitor motor with thermal overload protection. At max speed of 1075 RPM, it delivers 2820 CFM and 1.25 inches static pressure. The fan has a baked powder coat finish and comes with mounting brackets. It is ETL-listed and ideal for commercial and industrial ventilation applications. The 16-inch diameter allows it to fit most standard wall sleeves.
Catalog No. 50-276-4817 Supplier Grainger Supplier No. 16WD02
The 16WD02 is a 16-inch diameter, 2-blade, direct drive, forward-curved centrifugal fan from Continental Fan Manufacturing. It is constructed of galvanized steel and has a 1/3 HP, 115V, single-phase, permanent split capacitor motor with thermal overload protection. At max speed of 1075 RPM, it delivers 2820 CFM and 1.25 inches static pressure. The fan has a baked powder coat finish and comes with mounting brackets. It is ETL-listed and ideal for commercial and industrial ventilation applications. The 16-inch diameter allows it to fit most standard wall sleeves.