Learn More
Grainger Bumper 3/8in. Black Rubber PK100
Catalog No. 502764369
Encompass_Preferred
The Grainger 16WC36 Bumper Door Stop is a door-mounted door stop made of rubber and steel. It has a diameter of 1-1/2", projects 2-1/4" from the door surface, and withstands up to 100 lbs of holding force. This door stop features a mounting bracket that attaches to the door's surface with two included screws. It has a black finish and is designed to quietly stop doors from hitting walls while protecting both the door and wall surface. This product is made in the USA and is commonly used in commercial buildings, schools, hospitals, and more.
Catalog No. 50-276-4369 Supplier Grainger Supplier No. 16WC36
The Grainger 16WC36 Bumper Door Stop is a door-mounted door stop made of rubber and steel. It has a diameter of 1-1/2", projects 2-1/4" from the door surface, and withstands up to 100 lbs of holding force. This door stop features a mounting bracket that attaches to the door's surface with two included screws. It has a black finish and is designed to quietly stop doors from hitting walls while protecting both the door and wall surface. This product is made in the USA and is commonly used in commercial buildings, schools, hospitals, and more.