Learn More
Grainger Bumper 5/8 in Black Rubber PK10
Catalog No. 502768087
Encompass_Preferred
The Grainger Approved Bumper is a 5/8 inch thick black rubber bumper that provides impact absorption and noise dampening. It has a durometer of 60A rubber and features beveled edges for easier installation. The bumpers have pre-drilled holes for mounting and come in a pack of 4. They can help protect equipment and absorb vibration in industrial settings. The bumpers measure 2 inches wide by 3 inches long.
Catalog No. 50-276-8087 Supplier Grainger Supplier No. 16YP35
The Grainger Approved Bumper is a 5/8 inch thick black rubber bumper that provides impact absorption and noise dampening. It has a durometer of 60A rubber and features beveled edges for easier installation. The bumpers have pre-drilled holes for mounting and come in a pack of 4. They can help protect equipment and absorb vibration in industrial settings. The bumpers measure 2 inches wide by 3 inches long.