Learn More
Grainger Bumper 5/8 in Black Rubber PK100
Catalog No. 502768021
Encompass_Preferred
The Grainger 16ZD75 is a Westward brand 1/4-inch drive socket set. It contains 15 SAE sockets ranging from 1/4 inch to 9/16 inch and 15 metric sockets ranging from 4mm to 14mm. The chrome vanadium sockets have a knurled design for oil and slip resistance. The set comes in a molded case for storage and portability. This socket set is ideal for automotive and machinery applications that require standard shallow sockets in both SAE and metric sizes. The sockets are made in Taiwan.
Catalog No. 50-276-8021 Supplier Grainger Supplier No. 16ZD75
The Grainger 16ZD75 is a Westward brand 1/4-inch drive socket set. It contains 15 SAE sockets ranging from 1/4 inch to 9/16 inch and 15 metric sockets ranging from 4mm to 14mm. The chrome vanadium sockets have a knurled design for oil and slip resistance. The set comes in a molded case for storage and portability. This socket set is ideal for automotive and machinery applications that require standard shallow sockets in both SAE and metric sizes. The sockets are made in Taiwan.