Learn More
Grainger Bumper 5/8 in Black Rubber PK25
Catalog No. 502768206
Encompass_Preferred
The Grainger Approved Bumper is a rubber bumper designed to absorb shock and reduce noise from impacts. It has a cylindrical shape with a 2 inch diameter and 1 inch thickness. The bumper is made from SBR rubber which provides good abrasion resistance. It can withstand temperatures from -20 to 180 degrees Fahrenheit. The bumpers feature beveled edges for easier installation and are sold individually. They are commonly used on material handling equipment, conveyors, and other industrial machinery to protect components from damage. The bumpers are black in color.
Catalog No. 50-276-8206 Supplier Grainger Supplier No. 16ZD37
The Grainger Approved Bumper is a rubber bumper designed to absorb shock and reduce noise from impacts. It has a cylindrical shape with a 2 inch diameter and 1 inch thickness. The bumper is made from SBR rubber which provides good abrasion resistance. It can withstand temperatures from -20 to 180 degrees Fahrenheit. The bumpers feature beveled edges for easier installation and are sold individually. They are commonly used on material handling equipment, conveyors, and other industrial machinery to protect components from damage. The bumpers are black in color.