Learn More
Grainger Bumper 5/8 in Black Rubber PK25
Catalog No. 502764325
Encompass_Preferred
The 16WD15 is a 16-inch diameter, 15-inch face width, double row, deep groove ball bearing from SKF. It has a dynamic load rating of 420 kN and a static load rating of 315 kN. The bearing is made of high carbon chromium steel and has a maximum speed rating of 2,100 rpm. It uses a steel cage and has an ABEC 1 precision rating. The 16WD15 requires no lubrication and is ideal for applications with heavy radial loads such as conveyor pulleys, crushers, vibrating screens, and mixer drives. This SKF bearing provides reliable performance in demanding industrial environments.
Catalog No. 50-276-4325 Supplier Grainger Supplier No. 16WD15
The 16WD15 is a 16-inch diameter, 15-inch face width, double row, deep groove ball bearing from SKF. It has a dynamic load rating of 420 kN and a static load rating of 315 kN. The bearing is made of high carbon chromium steel and has a maximum speed rating of 2,100 rpm. It uses a steel cage and has an ABEC 1 precision rating. The 16WD15 requires no lubrication and is ideal for applications with heavy radial loads such as conveyor pulleys, crushers, vibrating screens, and mixer drives. This SKF bearing provides reliable performance in demanding industrial environments.