Learn More
Grainger Bumper 5/8 in Black Rubber PK25
Catalog No. 502763766
Encompass_Preferred
The Grainger Approved 16WD17 Bumper is a black rubber bumper with a 2-1/2" projection. It has a 1-1/2" diameter threaded stud mount and is rated for temperatures from -20°F to +180°F. This bumper absorbs shock and protects equipment and components from impact damage. It is made from oil and water resistant rubber. The bumper has a tensile strength of 2000 psi and a durometer hardness of 60 Shore A. It meets military specification MIL-B-15019F. The 16WD17 model weighs 0.1 lbs and has overall dimensions of 2-1/2" x 2" x 2-1/2". This industrial grade bumper provides impact absorption for equipment in factories, warehouses, and other commercial facilities.
Catalog No. 50-276-3766 Supplier Grainger Supplier No. 16WD17
The Grainger Approved 16WD17 Bumper is a black rubber bumper with a 2-1/2" projection. It has a 1-1/2" diameter threaded stud mount and is rated for temperatures from -20°F to +180°F. This bumper absorbs shock and protects equipment and components from impact damage. It is made from oil and water resistant rubber. The bumper has a tensile strength of 2000 psi and a durometer hardness of 60 Shore A. It meets military specification MIL-B-15019F. The 16WD17 model weighs 0.1 lbs and has overall dimensions of 2-1/2" x 2" x 2-1/2". This industrial grade bumper provides impact absorption for equipment in factories, warehouses, and other commercial facilities.