Learn More
Grainger Bumper 5/8in. Black Rubber PK10
Catalog No. 502768213
Encompass_Preferred
The Grainger 16YP38 is a Yale YP38-10 3/8" drive click-type torque wrench with a range of 10 to 150 ft-lb. It features a reversible ratchet head, knurled grip, and a positive lock with torque adjustment knob. This click-type wrench emits an audible click when the preset torque value is reached. It has an accuracy of ±4% clockwise and includes a blow molded case for storage and protection. The torque wrench has a length of 18.5" and weighs 3.4 lbs. It is ideal for precise tightening applications.
Catalog No. 50-276-8213 Supplier Grainger Supplier No. 16YP38
The Grainger 16YP38 is a Yale YP38-10 3/8" drive click-type torque wrench with a range of 10 to 150 ft-lb. It features a reversible ratchet head, knurled grip, and a positive lock with torque adjustment knob. This click-type wrench emits an audible click when the preset torque value is reached. It has an accuracy of ±4% clockwise and includes a blow molded case for storage and protection. The torque wrench has a length of 18.5" and weighs 3.4 lbs. It is ideal for precise tightening applications.