Learn More
Grainger Bumper 5/8in. Black Rubber PK25
Catalog No. 502764396
Encompass_Preferred
The Grainger Approved 16WH28 Bumper is a black rubber bumper with a 2-1/2" projection and a 1-1/2" hole diameter. It is made of SBR rubber and has a durometer of 60A. This bumper provides impact absorption and has applications including material handling equipment, conveyors, and dock equipment. It measures 2-1/2" x 2-1/2" x 1-1/2" and weighs 0.1 lbs. The operating temperature range is -20°F to 180°F.
Catalog No. 50-276-4396 Supplier Grainger Supplier No. 16WH28
The Grainger Approved 16WH28 Bumper is a black rubber bumper with a 2-1/2" projection and a 1-1/2" hole diameter. It is made of SBR rubber and has a durometer of 60A. This bumper provides impact absorption and has applications including material handling equipment, conveyors, and dock equipment. It measures 2-1/2" x 2-1/2" x 1-1/2" and weighs 0.1 lbs. The operating temperature range is -20°F to 180°F.