Learn More
Grainger Bumper 9/16 in Black Rubber PK25
Catalog No. 502768170
Encompass_Preferred
The Grainger 9-16 in. Black Rubber Bumper is a rubber bumper that provides impact absorption and noise reduction. It has a 9 inch outside diameter, 16 inch inside diameter, and is 2 inches thick. The bumper is made of black virgin SBR rubber with a durometer of 60A. It comes in a pack of 25 bumpers. The bumpers can help protect equipment and absorb vibration in industrial applications. Their solid rubber construction makes them durable and long-lasting.
Catalog No. 50-276-8170 Supplier Grainger Supplier No. 16YW60
The Grainger 9-16 in. Black Rubber Bumper is a rubber bumper that provides impact absorption and noise reduction. It has a 9 inch outside diameter, 16 inch inside diameter, and is 2 inches thick. The bumper is made of black virgin SBR rubber with a durometer of 60A. It comes in a pack of 25 bumpers. The bumpers can help protect equipment and absorb vibration in industrial applications. Their solid rubber construction makes them durable and long-lasting.