Learn More
Vernier™ Bumper and Launcher Kit
SDPSCI_ED
Catalog No. S15969ND Shop All Vernier Software Products
Change view
Click to view available options
Product Type: Bumper and Launcher Kit
Catalog No.
S15969ND Product Type: Bumper and Launcher Kit
1 options
Catalog No. S15969ND Supplier Vernier™ Supplier No. BLK
Add to Cart
Add to Cart
Includes
Bumper and Launcher Kit
The Bumper and Launcher Kit contains accessories for experiments involving the Vernier Dual-Range Force Sensor, Vernier Wireless Dynamics System and the Vernier Dynamics System. The accessories allow you to perform a wide range of experiments in kinematics and dynamics.
  • Contains accessories for experiments involving the Dual-Range Force Sensor, Wireless Dynamics Sensor System, and Dymanics Cart and Track System
  • Includes:
    • One track bracket
    • One dual-magnet bumper
    • One DFS end mount screw
    • Two magnetic bumpers
    • Two rubber bumpers
    • Two hoop bumpers
    • Two clay holders
    • Clay
Warranty and Services
5-year limited warranty
Specifications
Product Type Kit, Bumper and Launcher
For Use With (Application) Educational use only
For Use With (Equipment) Dual-Range Force Sensor, Acceleration Sensor, Dynamics Cart and Track System, Motion Encoder System