Learn More
Teco Diagnostics BUN REAGENT SET
Catalog No. NC9018987
Encompass_Preferred
For the direct colorimetric determination of Urea Nitrogen (BUN) in human serum or plasma. 2x100mL Vials of BUN Enzyme Reagent (Color), 2x100mL Vials of BUN Color Developer, 1x5mL Vial of BUN Standard,
Catalog No. NC9018987 Supplier Teco Diagnostics Supplier No. B551132
For the direct colorimetric determination of Urea Nitrogen (BUN) in human serum or plasma. 2x100mL Vials of BUN Enzyme Reagent (Color), 2x100mL Vials of BUN Color Developer, 1x5mL Vial of BUN Standard,