Learn More
MSC Value Collection ZMSCH70008 3/16" ID x 5/16" OD Dash 008 Buna-N O-Ring 1/16" Thick, Round Cross Section, Shore 65 - 75, -30 to 250°F
Catalog No. NC1673189
Encompass
Value Collection ZMSCH70008 3/16" ID x 5/16" OD Dash 008 Buna-N O-Ring 1/16" Thick, Round Cross Section, Shore 65 - 75, -30 to 250°F
Catalog No. NC1673189 Supplier MSC Supplier No. 09260084
Value Collection ZMSCH70008 3/16" ID x 5/16" OD Dash 008 Buna-N O-Ring 1/16" Thick, Round Cross Section, Shore 65 - 75, -30 to 250°F