Learn More
MSC 7.6Mm Id X 12.4Mm Od Buna-N O-Ring
Catalog No. NC1751022
Encompass
2.4mm Thick, Round Cross Section, Shore 65 - 75, -30 to 250°F
Catalog No. NC1751022 Supplier MSC Supplier No. 31958465
2.4mm Thick, Round Cross Section, Shore 65 - 75, -30 to 250°F