Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bunamidine hydrochloride | 1055-55-6 | 99.8% | C25H39ClN2O | 10 MM/1 ML
SDP

Catalog No. 5000355740
Encompass_Preferred

Bunamidine hydrochloride is a veterinary anti-platyhelmintic agent effective against *Echinococcus granulosus* and *Taenia hydatigena*. This product is supplied as a 10 mM solution in 1 mL, strictly for research applications.

  • Veterinary anti-platyhelmintic agent
  • Targets *Echinococcus granulosus* and *Taenia hydatigena*
  • For research use only
  • Purity: 99.8%
  • Chemical formula: C25H39ClN2O
  • CAS number: 1055-55-6
  • Recommended storage (in solvent): -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light

Catalog No. 50-003-55740 Supplier Medchemexpress LLC Supplier No. HY14507910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Bunamidine hydrochloride is a veterinary anti-platyhelmintic agent effective against *Echinococcus granulosus* and *Taenia hydatigena*. This product is supplied as a 10 mM solution in 1 mL, strictly for research applications.

  • Veterinary anti-platyhelmintic agent
  • Targets *Echinococcus granulosus* and *Taenia hydatigena*
  • For research use only
  • Purity: 99.8%
  • Chemical formula: C25H39ClN2O
  • CAS number: 1055-55-6
  • Recommended storage (in solvent): -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.