Learn More
Medchemexpress LLC Bunamidine (hydrochloride) | 1055-55-6 | 99.8% | C25H39ClN2O | 25 MG
SDP
Catalog No. 5000357169
Encompass_Preferred
Bunamidine hydrochloride is a veterinary anti-platyhelmintic agent primarily used for *Echinococcus granulosus* and *Taenia hydatigena*. This compound has a molecular weight of 419.04 and appears as a white to light yellow solid. It is intended for research use only and not for patient applications.
  • Veterinary anti-platyhelmintic agent
  • Used for Echinococcus granulosus and Taenia hydatigena
  • White to light yellow solid appearance
  • Research use only
Catalog No. 50-003-57169 Supplier Medchemexpress LLC Supplier No. 25MGHY145079
Bunamidine hydrochloride is a veterinary anti-platyhelmintic agent primarily used for *Echinococcus granulosus* and *Taenia hydatigena*. This compound has a molecular weight of 419.04 and appears as a white to light yellow solid. It is intended for research use only and not for patient applications.
  • Veterinary anti-platyhelmintic agent
  • Used for Echinococcus granulosus and Taenia hydatigena
  • White to light yellow solid appearance
  • Research use only