Learn More
Medchemexpress LLC α-Bungarotoxin | 11032-79-4 | 99.0% | 7984.12 | 1 MG
SDP
Catalog No. 5000422334
Encompass_Preferred
α-Bungarotoxin is a potent competitive antagonist at nicotinic acetylcholine receptors (nAChRs). It acts as a selective α7 receptor blocker, inhibiting α7 currents with an IC50 of 1.6 nM while showing no effect on α3β4 currents at concentrations up to 3 μM. This toxin binds specifically and with high affinity to the nicotinic acetylcholine receptor, competing with natural ligands and acting as a powerful inhibitor of its function, resulting in functional blockade of AcChoRs at the neuromuscular junction.
  • Competitive antagonist at nicotinic acetylcholine receptors
  • Selective α7 receptor blocker
  • Inhibits α7 currents with an IC50 of 1.6 nM
  • Binds specifically and with high affinity to the nicotinic acetylcholine receptor
  • Competes with natural ligand binding
  • Potent competitive inhibitor of nicotinic acetylcholine receptor function
Catalog No. 50-004-22334 Supplier Medchemexpress LLC Supplier No. HYP12641MG
α-Bungarotoxin is a potent competitive antagonist at nicotinic acetylcholine receptors (nAChRs). It acts as a selective α7 receptor blocker, inhibiting α7 currents with an IC50 of 1.6 nM while showing no effect on α3β4 currents at concentrations up to 3 μM. This toxin binds specifically and with high affinity to the nicotinic acetylcholine receptor, competing with natural ligands and acting as a powerful inhibitor of its function, resulting in functional blockade of AcChoRs at the neuromuscular junction.
  • Competitive antagonist at nicotinic acetylcholine receptors
  • Selective α7 receptor blocker
  • Inhibits α7 currents with an IC50 of 1.6 nM
  • Binds specifically and with high affinity to the nicotinic acetylcholine receptor
  • Competes with natural ligand binding
  • Potent competitive inhibitor of nicotinic acetylcholine receptor function