Learn More
Alomone Labs, LTD α-Bungarotoxin-Biotin (#B-100-B) 10 µg, is a highly pure, natural, and biologically active conjugated peptide toxin.
Catalog No. NC3999164
Encompass
Long neurotoxin 1, α-Bgtx, α-BuTXA Biotinylated Conjugate for Sensitive Detection of Neuromuscular Junctions, GABA(A) Receptors, nAChRs, and α-Bungarotoxin Binding Sites
Catalog No. NC3999164 Supplier Alomone Labs, LTD Supplier No. B100B10UG
Long neurotoxin 1, α-Bgtx, α-BuTXA Biotinylated Conjugate for Sensitive Detection of Neuromuscular Junctions, GABA(A) Receptors, nAChRs, and α-Bungarotoxin Binding Sites