Learn More
Enterprise Technology Solutions Bunn 3-Warmer Brewer,12-Cup,16"x21-1/5"x16-4/5",Stainless/Steel
SDP
Catalog No. 5000810492
Encompass_Preferred
12-cup Digital Three-Warmer Commercial Brewer offers a large 200 oz. tank (5.9 liters) for back-to-back brewing capacity. Brewer adjusts automatically to varying water pressure. For high lime areas, BrewLOGIC technology calculates flow rate and adjusts brew time to maintain consistent pot levels. BrewWIZARD technology incorporates simplified programming with a LCD screen. Pot level, cold brew lockout and tank temperature are set easily from advanced touch pad on front of machine. SplashGard funnel deflects hot liquids away from your hand. Coffee extraction is controlled with programmable pre-infusion or pulse brew and digital temperature control. Brewer offers digital temperature control and accuracy, automatic warmer shutoff, and dual voltage adaptability. Brewer can operate at 120V/15 amp or 120/208-240V/20 amp. Decanters are sold separately. Brewer is UL listed for commercial and industrial use. Brewer must be connected to a cold water system.
Catalog No. 50-008-10492 Supplier Enterprise Technology Solutions Supplier No. BUN387000002
12-cup Digital Three-Warmer Commercial Brewer offers a large 200 oz. tank (5.9 liters) for back-to-back brewing capacity. Brewer adjusts automatically to varying water pressure. For high lime areas, BrewLOGIC technology calculates flow rate and adjusts brew time to maintain consistent pot levels. BrewWIZARD technology incorporates simplified programming with a LCD screen. Pot level, cold brew lockout and tank temperature are set easily from advanced touch pad on front of machine. SplashGard funnel deflects hot liquids away from your hand. Coffee extraction is controlled with programmable pre-infusion or pulse brew and digital temperature control. Brewer offers digital temperature control and accuracy, automatic warmer shutoff, and dual voltage adaptability. Brewer can operate at 120V/15 amp or 120/208-240V/20 amp. Decanters are sold separately. Brewer is UL listed for commercial and industrial use. Brewer must be connected to a cold water system.