Learn More
Andwin Scientific BUNN CW15 APS COMM BREWER 120V
SDP
Catalog No. NC3786082
Encompass
Brews directly into 1.9 to 3 liter airpotsModels are space saving 9in wideAll stainless steel constructionPourover brewer requires no plumbing – completely portableSplashGard funnel deflects hot liquids away from the handAccommodates 1.9 to 3L airpotsAirpots are easily transported to remote meeting rooms/ breakfast bars/ etcAirpot not included
Catalog No. NC3786082 Supplier Andwin Scientific Supplier No. B000FAMS50
Brews directly into 1.9 to 3 liter airpotsModels are space saving 9in wideAll stainless steel constructionPourover brewer requires no plumbing – completely portableSplashGard funnel deflects hot liquids away from the handAccommodates 1.9 to 3L airpotsAirpots are easily transported to remote meeting rooms/ breakfast bars/ etcAirpot not included