Learn More
Zymo Research Corporation Bunny Wipe Fecal Collector
SDP
Catalog No. 502865901
Encompass_Preferred
The Bunny Wipe fecal sample collector is a user-friendly device designed for convenient at-home fecal collection for nucleic acid analysis. It replaces traditional collection methods like cups or scoops with a familiar toilet-paper-like design, offering a more comfortable and less messy experience. The Bunny Wipe works in tandem with the DNA/RNA Shield Fecal Collection Tube (R1101), which stabilizes the sample using DNA/RNA Shield reagent, inactivating viruses for safe transport. Samples stored in these tubes remain stable at ambient temperature and can be frozen for longer storage. To use, simply collect the sample with the Bunny Wipe, peel off the strip containing the material, and store it in the collection tube. This system is ideal for at-home fecal collection for nucleic acid extraction and analysis. Zymo Research also offers customizable collection kits for specific applications.
Catalog No. 50-286-5901 Supplier Zymo Research Corporation Supplier No. R113310
The Bunny Wipe fecal sample collector is a user-friendly device designed for convenient at-home fecal collection for nucleic acid analysis. It replaces traditional collection methods like cups or scoops with a familiar toilet-paper-like design, offering a more comfortable and less messy experience. The Bunny Wipe works in tandem with the DNA/RNA Shield Fecal Collection Tube (R1101), which stabilizes the sample using DNA/RNA Shield reagent, inactivating viruses for safe transport. Samples stored in these tubes remain stable at ambient temperature and can be frozen for longer storage. To use, simply collect the sample with the Bunny Wipe, peel off the strip containing the material, and store it in the collection tube. This system is ideal for at-home fecal collection for nucleic acid extraction and analysis. Zymo Research also offers customizable collection kits for specific applications.