Learn More
Medchemexpress LLC Buparlisib | 944396-07-0 | 99.8% | C18H21F3N6O2 | 200 MG
SDP
Catalog No. 5000434615
Encompass_Preferred
Buparlisib is a pan-class I PI3K inhibitor that exhibits blood-brain barrier permeability.
  • Pan-class I PI3K inhibitor
  • Blood-brain barrier permeable
  • Induces apoptosis in multiple myeloma cells
  • Causes dose-dependent growth inhibition in tested multiple myeloma cell lines
Catalog No. 50-004-34615 Supplier Medchemexpress LLC Supplier No. HY70063200MG
Buparlisib is a pan-class I PI3K inhibitor that exhibits blood-brain barrier permeability.
  • Pan-class I PI3K inhibitor
  • Blood-brain barrier permeable
  • Induces apoptosis in multiple myeloma cells
  • Causes dose-dependent growth inhibition in tested multiple myeloma cell lines