Learn More
Selleck Chemical LLC Buparlisib BKM120-5mg
SDP
Supplier:  Selleck Chemical LLC S22475MG
Encompass_Preferred
Buparlisib (BKM120, NVP-BKM120) is a selective PI3K inhibitor of p110α/β/δ/γ with IC50 of 52 nM/166 nM/116 nM/262 nM in cell-free assays, respectively. Reduced potency against VPS34, mTOR, DNAPK, with little activity to PI4Kβ. Buparlisib induces apoptosis. Phase 2.
Catalog No. 50-238-9904