Learn More
Medchemexpress LLC Buparlisib Hydrochloride | 1312445-63-8 | C18H22ClF3N6O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1518010MM/1ML
Encompass_Preferred
Buparlisib Hydrochloride (BKM120 Hydrochloride) is a CNS-penetrant pan-class I PI3K inhibitor with IC50 values of 52 nM for p110α, 166 nM for p110β, 116 nM for p110δ, and 262 nM for p110γ. It induces dose- and time-dependent apoptosis in multiple myeloma cell lines.
  • Active against class I PI3K's, including common p110α mutants
  • Lower potency against class III and IV PI3K's
  • Induces dose-dependent growth inhibition in multiple myeloma cell lines
  • Oral administration leads to dose-dependent modulation of pAKTSer473 in xenograft tumors
Catalog No. 50-003-74180