Learn More
Cayman Chemical Buparvaquon 10mg
SDP
Catalog No. 502987639
Encompass_Preferred
An inhibitor of cytochrome bc1 in the parasites T. annulata and T. parva (in vitro EC50 of 1.5 x 10-8 M and 6.1 x 10-10 M, respectively)
Catalog No. 50-298-7639 Supplier Cayman Chemical Supplier No. 2170410 MG
An inhibitor of cytochrome bc1 in the parasites T. annulata and T. parva (in vitro EC50 of 1.5 x 10-8 M and 6.1 x 10-10 M, respectively)