Learn More
Medchemexpress LLC HY-17581 10mg Medchemexpress, Buparvaquone CAS:88426-33-9 Purity:>98%
SDP
Catalog No. 502081992
Encompass_Preferred
Medchemexpress, HY-17581 10mg Buparvaquone CAS:88426-33-9 Buparvaquone is a hydroxynaphthoquinone antiprotozoal drug related to parvaquone and atovaquone. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1992 Supplier Medchemexpress LLC Supplier No. HY1758110MG
Medchemexpress, HY-17581 10mg Buparvaquone CAS:88426-33-9 Buparvaquone is a hydroxynaphthoquinone antiprotozoal drug related to parvaquone and atovaquone. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.