Learn More
Apexbio Technology LLC Buparvaquone, 10mg CAS# 88426-33-9
SDP
Supplier:  Apexbio Technology LLC B472510
Encompass_Preferred
Buparvaquone is an analog of the antimalarial hydroxynaphthoquinones. It is applicable for the therapy and prophylaxis of Theileriosis and exhibits an anti-Leishmanial activity. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8242