Learn More
Cayman Chemical Buphedron metabolItehydroc
SDP
Catalog No. 502893175
Encompass_Preferred
A metabolite of buphedrone that features conversion of the β-keto group to β-hydroxy and is an enantiomeric mixture of the R,S and S,R orientations at carbons one and two, as in ephedrine
Catalog No. 50-289-3175 Supplier Cayman Chemical Supplier No. 9001432500 UG
A metabolite of buphedrone that features conversion of the β-keto group to β-hydroxy and is an enantiomeric mixture of the R,S and S,R orientations at carbons one and two, as in ephedrine