Learn More
Cayman Chemical BupIvacaIn-d9 10mg
SDP
Catalog No. 502965808
Encompass_Preferred
An internal standard for the quantification of bupivacaine by GC- or LC-MS
Catalog No. 50-296-5808 Supplier Cayman Chemical Supplier No. 1824610 MG
An internal standard for the quantification of bupivacaine by GC- or LC-MS