Learn More
Lkt Laboratories Bupivacaine, ≥98%, 5GR
SDP
Supplier:  Lkt Laboratories B82615G
Encompass_Preferred
BK/SK, Kv1, Kv3, TASK-2 K+ channel blocker, voltage-gated Na+ channel blocker. CAS: 38396-39-3. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4872