Learn More
Apexbio Technology LLC Bupivacaine HCl 18010-40-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062068
Encompass_Preferred
Bupivacaine Hydrochloride (CAS 18010-40-7) is an amino amide-type local anesthetic compound that functions primarily through blockade of voltage-gated sodium channels By preventing sodium ion influx in neuronal membranes it reduces action potential propagation resulting in reversible inhibition of nerve conduction Due to its relatively rapid onset paired with prolonged anesthetic duration bupivacaine hydrochloride is frequently utilized experimentally to investigate neuronal signaling pain mechanisms and local anesthetic pharmacology in biomedical research
Catalog No. 50-000-62068 Supplier Apexbio Technology LLC Supplier No. B14205.1
Bupivacaine Hydrochloride (CAS 18010-40-7) is an amino amide-type local anesthetic compound that functions primarily through blockade of voltage-gated sodium channels By preventing sodium ion influx in neuronal membranes it reduces action potential propagation resulting in reversible inhibition of nerve conduction Due to its relatively rapid onset paired with prolonged anesthetic duration bupivacaine hydrochloride is frequently utilized experimentally to investigate neuronal signaling pain mechanisms and local anesthetic pharmacology in biomedical research