Learn More
Medchemexpress LLC HY-B0405A 100mg Medchemexpress, Bupivacaine (hydrochloride) CAS:18010-40-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0405A100MG
Encompass_Preferred
Medchemexpress, HY-B0405A 100mg Bupivacaine (hydrochloride) CAS:18010-40-7 Bupivacaine hydrochloride is a Na+ channel blocker which has local narcotic effect. Bupivacaine hydrochloride has direct anti-cancer activities with the dominant inhibitory effects on gastric cancer migration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2523
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.