Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bupivacaine Hydrochloride, 100.3%, MP Biomedicals™

Catalog No. ICN15490405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15490405 5 g
ICN15490401 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15490405 Supplier MP Biomedicals, Inc Supplier No. 0215490405
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14252-80-3
Molecular Formula C18H29ClN2O
Molecular Weight (g/mol) 324.89
MDL Number MFCD00078956
InChI Key SIEYLFHKZGLBNX-UHFFFAOYNA-N
Synonym bupivacaine hydrochloride, bupivacaine hcl, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrochloride, vivacaine, marcain, +--bupivacaine hydrochloride, bupivacaine hydrochloride jan, 1-butyl-n-2,6-dimethylphenyl-2-piperidinecarboxamide monohydrochloride, 2',6'-pipecoloxylidide, 1-butyl-, hydrochloride
PubChem CID 64737
ChEBI CHEBI:77441
IUPAC Name hydrogen 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide chloride
SMILES [H+].[Cl-].CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C

Specifications

Color White
Assay Percent Range 100.3%
Quantity 5 g
Formula Weight 324.9 (anhydrous)
Physical Form Powder
Chemical Name or Material Bupivacaine Hydrochloride
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.