Learn More
Medchemexpress LLC Bupivacaine (hydrochloride) | 18010-40-7 | 100.0% | 50 MG
Supplier: Medchemexpress LLC HYB0405AR50MG
Bupivacaine hydrochloride (Standard) is an analytical standard intended for research and analytical applications. It acts as an NMDA receptor inhibitor and can block sodium, L-calcium, and potassium channels. Potently blocking SCN5A channels with an IC50 of 69.5 μM, it can be used for chronic pain research.
- Analytical standard for research and analytical applications.
- NMDA receptor inhibitor.
- Blocks sodium, L-calcium, and potassium channels.
- Potently blocks SCN5A channels with an IC50 of 69.5 μM.
- Useful for chronic pain research.
- Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.